siRNA Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284392.1 | NP_001271321.1 | 9, 10 | 1079 | ||
| NM_011129.2 | NP_035259.1 | 10, 11 | 1376 | |||
| GenBank mRNA | AB078010.1 | - | 9, 10 | 974 | ||
| AK135143.1 | - | 10, 11 | 1377 | |||
| X61452.1 | - | 10, 11 | 1306 | |||

Sept4
Genomic Map