siRNA Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284392.1 | NP_001271321.1 | 6 | 713 | ||
| NM_001284394.1 | NP_001271323.1 | 7 | 1010 | |||
| NM_001284398.1 | NP_001271327.1 | 6 | 697 | |||
| NM_011129.2 | NP_035259.1 | 7 | 1010 | |||
| GenBank mRNA | AB078010.1 | - | 6 | 608 | ||
| AK133933.1 | - | 6 | 665 | |||
| AK135143.1 | - | 7 | 1011 | |||
| BC055101.1 | - | 7 | 1010 | 6591016 | ||
| X61452.1 | - | 7 | 940 | |||

Sept4
Genomic Map