siRNA Details
| Gene Symbol: Polg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017462.2 | NP_059490.2 | 4 | 1183 | ||
| GenBank mRNA | AB121698.1 | - | 3 | 876 | ||
| AK133584.1 | - | 4 | 1109 | |||
| AK148091.1 | - | 4 | 1145 | |||
| BC053024.1 | - | 4 | 1111 | |||
| BC061472.1 | - | 4 | 1097 | 6827238 | ||
| BC080291.1 | - | 4 | 1090 | |||
| U53584.1 | - | 4 | 1103 | |||

Polg
Genomic Map