siRNA Details
| Gene Symbol: Pou3f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008901.1 | NP_032927.1 | 1 | 983 | ||
| GenBank mRNA | BC052012.1 | - | 1 | 1031 | ||
| BC086670.1 | - | 1046 | ||||
| BC138657.1 | - | 1 | 1058 | 8861677 | ||
| BC145919.1 | - | 1 | 1058 | 40131222 | ||
| X66603.1 | - | 1026 | ||||

Pou3f4
Genomic Map