siRNA Details
| Gene Symbol: Ctsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038492.2 | NP_001033581.1 | 13 | 1523 | ||
| NM_008906.4 | NP_032932.2 | 13 | 1276 | |||
| GenBank mRNA | AK019861.1 | - | 13 | 1481 | ||
| AK032197.1 | - | 13 | 1243 | |||
| AK159150.1 | - | 13 | 1244 | |||
| AK159766.1 | - | 13 | 1239 | |||
| AK196352.1 | - | 2 | 103 | |||
| BC018534.1 | - | 13 | 1232 | 2651479 | ||
| J05261.1 | - | 13 | 1411 | |||

Ctsa
Genomic Map