siRNA Details
| Gene Symbol: Ppp1r17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011153.3 | NP_035283.1 | 5 | 559 | ||
| GenBank mRNA | AF071562.1 | - | 5 | 557 | ||
| AF500906.1 | - | 5 | 569 | |||
| AK078335.1 | - | 5 | 576 | |||
| BC026822.1 | - | 5 | 559 | 4511839 | ||

Ppp1r17
Genomic Map