siRNA Details
| Gene Symbol: Ppp3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293622.1 | NP_001280551.1 | 4 | 1254 | ||
| NM_008913.5 | NP_032939.1 | 4 | 1254 | |||
| GenBank mRNA | AK146387.1 | - | 4 | 1198 | ||
| AK150393.1 | - | 4 | 613 | |||
| AK166287.1 | - | 4 | 1031 | |||
| BC138612.1 | - | 4 | 527 | 8861632 | ||
| J05479.1 | - | 4 | 551 | |||

Ppp3ca
Genomic Map