siRNA Details
| Gene Symbol: Ppp3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024459.2 | NP_077779.2 | 4 | 499 | ||
| GenBank mRNA | AK133403.1 | - | 470 | |||
| AK145541.1 | - | 4 | 476 | |||
| AK160423.1 | - | 444 | ||||
| AK160938.1 | - | 4 | 485 | |||
| AK163241.1 | - | 4 | 473 | |||
| BC002267.1 | - | 2 | 327 | |||
| S43864.1 | - | 3 | 257 | |||

Ppp3r1
Genomic Map