siRNA Details
Gene Symbol: Nup88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083331.2 | NP_001076800.1 | 10 | 1476 | ||
NM_001276406.1 | NP_001263335.1 | 10 | 1476 | |||
NM_172394.3 | NP_765982.2 | 10 | 1476 | |||
GenBank mRNA | AJ532593.1 | - | 10 | 1470 | ||
AK002643.1 | - | 10 | 1462 | |||
AK028563.1 | - | 10 | 1466 | |||
AK050893.1 | - | 10 | 1465 | |||
AK051122.1 | - | 1 | 1081 | |||
AK085776.1 | - | 10 | 1475 | |||
AK139738.1 | - | 9 | 1843 | |||
AK160619.1 | - | 10 | 1477 | |||
AK166214.1 | - | 10 | 1466 | |||
BC032929.1 | - | 10 | 1457 | 1379673 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053616, BC072524, U93692.
Panther Classification:
Gene Ontology Categories:
