siRNA Details
| Gene Symbol: Prkar2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008924.2 | NP_032950.1 | 6 | 878 | ||
| GenBank mRNA | AF533977.1 | - | 6 | 878 | ||
| AK132728.1 | - | 6 | 817 | |||
| AK132824.1 | - | 6 | 809 | |||
| BC075623.1 | - | 6 | 849 | 6854057 | ||
| BC080276.1 | - | 6 | 872 | 30354403 | ||
| J02935.1 | - | 6 | 561 | |||

Prkar2a
Genomic Map