siRNA Details
| Gene Symbol: Prl4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011165.3 | NP_035295.1 | 3 | 287 | ||
| GenBank mRNA | AF011383.1 | - | 3 | 288 | ||
| AF015562.1 | - | 3 | 287 | |||
| AK141285.1 | - | 3 | 299 | |||
| AK164009.1 | - | 3 | 299 | |||
| BC052218.1 | - | 3 | 288 | 30017285 | ||

Prl4a1
Genomic Map