siRNA Details
| Gene Symbol: Prl6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011166.2 | NP_035296.1 | 3 | 259 | ||
| GenBank mRNA | AF011384.1 | - | 3 | 258 | ||
| AF015563.1 | - | 3 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl6a1| Gene Symbol: Prl6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011166.2 | NP_035296.1 | 3 | 259 | ||
| GenBank mRNA | AF011384.1 | - | 3 | 258 | ||
| AF015563.1 | - | 3 | 259 | |||