siRNA Details
| Gene Symbol: Prl7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164058.1 | NP_001157530.1 | 2 | 145 | ||
| NM_008930.2 | NP_032956.1 | 2 | 142 | |||
| GenBank mRNA | AF011381.1 | - | 2 | 134 | ||
| AF020525.1 | - | 2 | 116 | |||
| AK028371.1 | - | 2 | 143 | |||
| AK141309.1 | - | 2 | 146 | |||
| AK146050.1 | - | 2 | 146 | |||
| BB611867.1 | - | 2 | 143 | |||
| BC051671.1 | - | 2 | 127 | 30018275 | ||

Prl7a1
Genomic Map