siRNA Details
| Gene Symbol: Prl7a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164058.1 | NP_001157530.1 | 6 | 549 | ||
| NM_008930.2 | NP_032956.1 | 6 | 546 | |||
| GenBank mRNA | AF011381.1 | - | 6 | 538 | ||
| AF020525.1 | - | 6 | 520 | |||
| AK028371.1 | - | 6 | 547 | |||
| AK141309.1 | - | 6 | 550 | |||
| AK146050.1 | - | 6 | 549 | |||
| BB611867.1 | - | 6 | 547 | |||
| BC051671.1 | - | 6 | 531 | 30018275 | ||

Prl7a1
Genomic Map