siRNA Details
| Gene Symbol: Prm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013638.2 | NP_038666.2 | 1 | 149 | ||
| GenBank mRNA | AK132894.1 | - | 1 | 150 | ||
| BC049598.1 | - | 140 | ||||
| BC061127.1 | - | 148 | 6771130 | |||
| BC090986.1 | - | 209 | 6740459 | |||

Prm3
Genomic Map