siRNA Details
| Gene Symbol: Prox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008937.2 | NP_032963.1 | 2 | 2050 | ||
| GenBank mRNA | AF061576.1 | - | 1 | 1637 | ||
| AK052889.1 | - | 2 | 2084 | |||
| AK143179.1 | - | 2 | 1874 | |||
| BC051411.1 | - | 2 | 2050 | 6490801 | ||

Prox1
Genomic Map