siRNA Details
| Gene Symbol: Prox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008937.2 | NP_032963.1 | 3, 4 | 2312 | ||
| GenBank mRNA | AF061576.1 | - | 2, 3 | 1899 | ||
| AK052889.1 | - | 3, 4 | 2346 | |||
| AK143179.1 | - | 3, 4 | 2136 | |||
| BC051411.1 | - | 3, 4 | 2312 | 6490801 | ||

Prox1
Genomic Map