siRNA Details
| Gene Symbol: Prph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008938.1 | NP_032964.1 | 1 | 367 | ||
| GenBank mRNA | AK044549.1 | - | 1 | 362 | ||
| AK136280.1 | - | 1 | 394 | |||
| BC028958.1 | - | 1 | 362 | 5363367 | ||
| BC085266.1 | - | 1 | 366 | 6490224 | ||
| X14770.1 | - | 1 | 367 | |||

Prph2
Genomic Map