siRNA Details
| Gene Symbol: Prss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008939.2 | NP_032965.1 | 9, 10 | 1942 | ||
| GenBank mRNA | AK132633.1 | - | 9, 10 | 1731 | ||
| AK186311.1 | - | 3, 4 | 271 | |||
| AK205149.1 | - | 2, 3 | 180 | |||
| BC031429.1 | - | 9, 10 | 1844 | 3665834 | ||
| D89871.1 | - | 9, 10 | 1940 | |||
| Y13192.1 | - | 9, 10 | 1719 | |||

Prss12
Genomic Map