siRNA Details
| Gene Symbol: IGFALS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146006.1 | NP_001139478.1 | 2 | 904 | ||
| NM_004970.2 | NP_004961.1 | 2 | 790 | |||
| NR_027389.1 | 2 | 763 | ||||
| GenBank mRNA | AK292182.1 | - | 2 | 757 | ||
| AK303146.1 | - | 2 | 903 | |||
| AK309598.1 | - | 2 | 725 | |||
| AK313198.1 | - | 2 | 757 | |||
| AK315808.1 | - | 2 | 783 | |||
| BC025681.1 | - | 2 | 790 | 5205012 | ||
| M86826.1 | - | 2 | 765 | |||

IGFALS
Genomic Map