siRNA Details
| Gene Symbol: Prss12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008939.2 | NP_032965.1 | 11 | 2266 | ||
| GenBank mRNA | AK132633.1 | - | 11 | 2055 | ||
| BC031429.1 | - | 11 | 2168 | 3665834 | ||
| D89871.1 | - | 11 | 2264 | |||
| Y13192.1 | - | 11 | 2043 | |||

Prss12
Genomic Map