siRNA Details
| Gene Symbol: Prx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019412.2 | NP_062285.1 | 6 | 2907 | ||
| NM_198048.2 | NP_932165.2 | 7 | 2315 | |||
| GenBank mRNA | AJ222968.1 | - | 7 | 2314 | ||
| AJ222969.1 | - | 6 | 2906 | |||
| AK132429.1 | - | 2 | 1964 | |||
| AK173220.1 | - | 3 | 2112 | |||
| BC068135.1 | - | 3 | 2128 | 30354483 | ||

Prx
Genomic Map