siRNA Details
| Gene Symbol: Cyth1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112699.1 | NP_001106169.1 | 13 | 1267 | ||
| NM_001112700.1 | NP_001106170.1 | 14 | 1210 | |||
| NM_011180.3 | NP_035310.2 | 14 | 1270 | |||
| GenBank mRNA | AB013464.1 | - | 13 | 1188 | ||
| AB035538.1 | - | 13 | 1239 | |||
| AF051337.1 | - | 13 | 1138 | |||
| AK158216.1 | - | 14 | 1210 | |||
| AK172296.1 | - | 14 | 1215 | |||
| AK220302.1 | - | 14 | 1192 | |||
| AY126516.1 | - | 13 | 1236 | |||
| BC057974.1 | - | 13 | 1151 | 5060913 | ||

Cyth1
Genomic Map