siRNA Details
| Gene Symbol: Cyth3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163548.1 | NP_001157020.1 | 2 | 600 | ||
| NM_011182.4 | NP_035312.3 | 2 | 276 | |||
| GenBank mRNA | AB013470.1 | - | 2 | 130 | ||
| AF084221.1 | - | 2 | 170 | |||
| AK016645.1 | - | 2 | 602 | |||
| AK036191.1 | - | 2 | 190 | |||
| AK142407.1 | - | 2 | 265 | |||
| AK159229.1 | - | 2 | 248 | |||
| AK168566.1 | - | 2 | 190 | |||
| AK220565.1 | - | 2 | 256 | |||
| BC007189.1 | - | 1 | 17 | |||
| BC035296.1 | - | 2 | 217 | 3969080 | ||

Cyth3
Genomic Map