siRNA Details
| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128605.1 | NP_001122077.1 | 3 | 403 | ||
| NM_011183.3 | NP_035313.2 | 3 | 412 | |||
| GenBank mRNA | AF038935.1 | - | 2 | 340 | ||
| AK014706.1 | - | 2 | 347 | |||
| AK148111.1 | - | 3 | 404 | |||
| AK154119.1 | - | 3 | 348 | |||
| BC010403.1 | - | 2 | 358 | 4038761 | ||
| U57324.1 | - | 3 | 363 | |||

Psen2
Genomic Map