siRNA Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008944.2 | NP_032970.2 | 2 | 95 | ||
| GenBank mRNA | AF099733.1 | - | 2 | 62 | ||
| AK012119.1 | - | 2 | 77 | |||
| AK012990.1 | - | 2 | 84 | |||
| AK035578.1 | - | 84 | ||||
| AK136115.1 | - | 2 | 97 | |||
| AK151883.1 | - | 2 | 86 | |||
| AK152862.1 | - | 2 | 86 | |||
| AK154011.1 | - | 2 | 79 | |||
| AK167752.1 | - | 2 | 86 | |||
| BC026768.1 | - | 2 | 68 | 3156907 | ||
| CJ052982.1 | - | 2 | 97 | |||
| X70303.1 | - | 2 | 139 | |||

Psma2
Genomic Map