siRNA Details
| Gene Symbol: Psmb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011185.3 | NP_035315.1 | 3 | 301 | ||
| GenBank mRNA | AK028265.1 | - | 3 | 322 | ||
| AK077520.1 | - | 3 | 302 | |||
| AK132161.1 | - | 3 | 302 | |||
| AK167647.1 | - | 3 | 323 | |||
| BC018351.1 | - | 3 | 302 | 3584538 | ||
| U60824.1 | - | 3 | 316 | |||
| X80686.1 | - | 3 | 305 | |||

Psmb1
Genomic Map