siRNA Details
| Gene Symbol: Psmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008945.3 | NP_032971.2 | 3 | 445 | ||
| GenBank mRNA | AK077448.1 | - | 3 | 422 | ||
| AK137044.1 | - | 3 | 421 | |||
| AK156624.1 | - | 3 | 421 | |||
| AK161348.1 | - | 3 | 421 | |||
| AK168265.1 | - | 3 | 421 | |||
| AK168340.1 | - | 3 | 421 | |||
| BC008241.1 | - | 3 | 400 | 2654421 | ||
| CB058553.1 | - | 3 | 445 | |||
| U65636.1 | - | 3 | 400 | |||

Psmb4
Genomic Map