siRNA Details
| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011186.1 | NP_035316.1 | 2 | 440 | ||
| GenBank mRNA | AF060091.1 | - | 2 | 440 | ||
| AK010441.1 | - | 2 | 460 | |||
| AK133839.1 | - | 2 | 461 | |||
| AK186367.1 | - | 1 | 91 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmb5| Gene Symbol: Psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011186.1 | NP_035316.1 | 2 | 440 | ||
| GenBank mRNA | AF060091.1 | - | 2 | 440 | ||
| AK010441.1 | - | 2 | 460 | |||
| AK133839.1 | - | 2 | 461 | |||
| AK186367.1 | - | 1 | 91 | |||