siRNA Details
| Gene Symbol: Psmb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008946.4 | NP_032972.3 | 4 | 405 | ||
| GenBank mRNA | AK078437.1 | - | 4 | 377 | ||
| AK132042.1 | - | 4 | 388 | |||
| AK167123.1 | - | 4 | 377 | |||
| AK167227.1 | - | 4 | 373 | |||
| BC013897.1 | - | 4 | 365 | 4038758 | ||
| U13393.1 | - | 4 | 326 | |||

Psmb6
Genomic Map