siRNA Details
| Gene Symbol: Ptch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008957.2 | NP_032983.1 | 22 | 3674 | ||
| GenBank mRNA | AK147626.1 | - | 23 | 3821 | ||
| AK163736.1 | - | 2 | 243 | |||
| U46155.1 | - | 22 | 3674 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptch1| Gene Symbol: Ptch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008957.2 | NP_032983.1 | 22 | 3674 | ||
| GenBank mRNA | AK147626.1 | - | 23 | 3821 | ||
| AK163736.1 | - | 2 | 243 | |||
| U46155.1 | - | 22 | 3674 | |||