siRNA Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008959.3 | NP_032985.2 | 4, 5 | 1127 | ||
| GenBank mRNA | AF042731.1 | - | 4, 5 | 542 | ||
| AK036990.1 | - | 4, 5 | 589 | |||
| AK054101.1 | - | 4, 5 | 566 | |||
| AK085857.1 | - | 4, 5 | 1128 | |||
| AK086136.1 | - | 3, 4 | 651 | |||
| AK088048.1 | - | 4, 5 | 609 | |||
| AK137681.1 | - | 4, 5 | 797 | |||
| AK172873.1 | - | 4, 5 | 1104 | |||
| AK187434.1 | - | 20 | ||||
| BC003260.1 | - | 4, 5 | 764 | 3501141 | ||

Ptdss1
Genomic Map