siRNA Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008959.3 | NP_032985.2 | 5 | 1169 | ||
| GenBank mRNA | AK036990.1 | - | 5 | 631 | ||
| AK054101.1 | - | 5 | 608 | |||
| AK085857.1 | - | 5 | 1170 | |||
| AK086136.1 | - | 4 | 693 | |||
| AK088048.1 | - | 5 | 651 | |||
| AK137681.1 | - | 5 | 839 | |||
| AK172873.1 | - | 5 | 1146 | |||
| BC003260.1 | - | 5 | 806 | 3501141 | ||

Ptdss1
Genomic Map