siRNA Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008959.3 | NP_032985.2 | 10 | 1775 | ||
| GenBank mRNA | AF042731.1 | - | 10 | 1190 | ||
| AK036990.1 | - | 10 | 1237 | |||
| AK054101.1 | - | 10 | 1214 | |||
| AK085857.1 | - | 10 | 1776 | |||
| AK086136.1 | - | 10 | 1434 | |||
| AK088048.1 | - | 10 | 1257 | |||
| AK137681.1 | - | 10 | 1445 | |||
| AK172873.1 | - | 9 | 3561 | |||
| BC003260.1 | - | 10 | 1412 | 3501141 | ||

Ptdss1
Genomic Map