siRNA Details
| Gene Symbol: Ptgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008963.2 | NP_032989.2 | 2 | 266 | ||
| GenBank mRNA | AB006361.1 | - | 2 | 249 | ||
| AK002382.1 | - | 1 | 91 | |||
| AK131859.1 | - | 2 | 266 | |||
| BC038083.1 | - | 1 | 342 | 4481308 | ||
| BC043015.1 | - | 2 | 252 | 5683706 | ||
| X89222.1 | - | 2 | 237 | |||

Ptgds
Genomic Map