siRNA Details
Gene Symbol: Ptger1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013641.2 | NP_038669.1 | 2 | 833 | ||
GenBank mRNA | AK053616.1 | - | 607 | |||
AK164746.1 | - | 641 | ||||
BC138739.1 | - | 2 | 795 | 8861761 | ||
BC138744.1 | - | 2 | 795 | 8861766 | ||
D16338.1 | - | 2 | 610 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
