siRNA Details
| Gene Symbol: Ptpn14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008976.2 | NP_033002.2 | 9, 10 | 1375 | ||
| GenBank mRNA | AF170902.1 | - | 8, 9 | 942 | ||
| AF170903.1 | - | 8, 9 | 942 | |||
| AK028783.1 | - | 9, 10 | 1339 | |||
| AK037646.1 | - | 9, 10 | 1376 | |||
| AK086524.1 | - | 9, 10 | 1369 | |||
| AK156480.1 | - | 9, 10 | 1121 | |||
| D31842.1 | - | 8, 9 | 972 | |||

Ptpn14
Genomic Map