siRNA Details
| Gene Symbol: Ptpn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011206.2 | NP_035336.2 | 14 | 1339 | ||
| GenBank mRNA | AK014043.1 | - | 276 | |||
| AK144353.1 | - | 14 | 1321 | |||
| U35124.1 | - | 14 | 1313 | |||
| U49853.1 | - | 14 | 1279 | |||
| U52523.1 | - | 14 | 1324 | |||

Ptpn18
Genomic Map