siRNA Details
| Gene Symbol: Ptpn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019933.2 | NP_064317.2 | 14 | 1285 | ||
| GenBank mRNA | AF106702.1 | - | 13 | 1113 | ||
| AK046683.1 | - | 14 | 1560 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptpn4| Gene Symbol: Ptpn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019933.2 | NP_064317.2 | 14 | 1285 | ||
| GenBank mRNA | AF106702.1 | - | 13 | 1113 | ||
| AK046683.1 | - | 14 | 1560 | |||