siRNA Details
| Gene Symbol: Ptpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163565.1 | NP_001157037.1 | 8 | 1246 | ||
| NM_013643.2 | NP_038671.2 | 8 | 1354 | |||
| GenBank mRNA | AK038416.1 | - | 8 | 1023 | ||
| BC051975.1 | - | 8 | 995 | |||
| BC057339.1 | - | 7 | 1516 | |||
| BC079592.1 | - | 8 | 1326 | 6817360 | ||
| S80329.1 | - | 5 | 1190 | |||
| U28216.1 | - | 8 | 1107 | |||
| U28217.1 | - | 8 | 1365 | |||

Ptpn5
Genomic Map