siRNA Details
| Gene Symbol: Ptprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029928.2 | NP_084204.2 | 24 | 5359 | ||
| GenBank mRNA | AF157628.1 | - | 8 | 852 | ||
| AK147439.1 | - | 24 | 5361 | |||
| AK147668.1 | - | 24 | 5361 | |||
| AK162366.1 | - | 4 | 256 | |||
| AY077755.1 | - | 24 | 5237 | |||
| BC047086.1 | - | 14 | 2419 | |||
| BC141006.1 | - | 24 | 5155 | 9055618 | ||
| BC145111.1 | - | 24 | 5155 | 9053659 | ||
| X58289.1 | - | 8 | 857 | |||

Ptprb
Genomic Map