siRNA Details
| Gene Symbol: Ptpre | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011212.3 | NP_035342.3 | 13 | 1356 | ||
| GenBank mRNA | AK004602.1 | - | 13 | 1358 | ||
| AK132455.1 | - | 12 | 1319 | |||
| AK154910.1 | - | 10 | 908 | |||
| AK160947.1 | - | 10 | 905 | |||
| BC053408.1 | - | 13 | 1132 | |||
| BC098473.1 | - | 13 | 1120 | |||
| D83484.1 | - | 13 | 1693 | |||
| U35368.1 | - | 13 | 1356 | |||
| U36758.1 | - | 10 | 905 | |||
| U40280.1 | - | 12 | 1096 | |||
| U62387.1 | - | 13 | 1258 | |||

Ptpre
Genomic Map