siRNA Details
| Gene Symbol: Ptprk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008983.2 | NP_033009.1 | 4, 5 | 830 | ||
| GenBank mRNA | AK019870.1 | - | 4, 5 | 730 | ||
| AK052275.1 | - | 4, 5 | 784 | |||
| AK083006.1 | - | 4, 5 | 794 | |||
| BC138376.1 | - | 4, 5 | 796 | 8861396 | ||
| BC145261.1 | - | 4, 5 | 796 | |||

Ptprk
Genomic Map