siRNA Details
| Gene Symbol: Ptpru | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083119.2 | NP_001076588.1 | 17, 18 | 2818 | ||
| NM_011214.2 | NP_035344.2 | 16, 17 | 2788 | |||
| GenBank mRNA | AK047640.1 | - | 16, 17 | 2516 | ||
| AK052720.1 | - | 16, 17 | 2763 | |||
| BC080736.1 | - | 16, 17 | 2743 | |||
| D88187.1 | - | 16, 17 | 2779 | |||

Ptpru
Genomic Map