siRNA Details
| Gene Symbol: Ptprn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011215.2 | NP_035345.2 | 9 | 1524 | ||
| GenBank mRNA | AK081267.1 | - | 9 | 1517 | ||
| AK138655.1 | - | 9 | 1516 | |||
| AK220255.1 | - | 9 | 1452 | |||
| BC133678.1 | - | 9 | 1408 | 40109882 | ||
| U57345.1 | - | 9 | 1411 | |||
| U82439.1 | - | 4 | 532 | |||

Ptprn2
Genomic Map