siRNA Details
| Gene Symbol: Purb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011221.3 | NP_035351.1 | 1 | 1087 | ||
| GenBank mRNA | AF017630.1 | - | 1 | 931 | ||
| AK049443.1 | - | 1 | 224 | |||
| AK139376.1 | - | 1 | 226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Purb| Gene Symbol: Purb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011221.3 | NP_035351.1 | 1 | 1087 | ||
| GenBank mRNA | AF017630.1 | - | 1 | 931 | ||
| AK049443.1 | - | 1 | 224 | |||
| AK139376.1 | - | 1 | 226 | |||