siRNA Details
| Gene Symbol: Abcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008991.2 | NP_033017.2 | 13 | 1238 | ||
| GenBank mRNA | AK004660.1 | - | 13 | 1219 | ||
| AK144060.1 | - | 13 | 1220 | |||
| AK153298.1 | - | 13 | 1196 | |||
| AK201522.1 | - | 4 | 322 | |||
| BC009119.1 | - | 13 | 1230 | 3154529 | ||
| BC054446.1 | - | 13 | 1186 | 6477711 | ||
| CB245413.1 | - | 8 | 686 | |||
| X89569.1 | - | 4 | 331 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012804, D90038.

Abcd3
Genomic Map