siRNA Details
| Gene Symbol: Pxn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011223.3 | NP_035353.1 | 10 | 1471 | ||
| NM_133915.3 | NP_598676.2 | 11 | 1573 | |||
| GenBank mRNA | AF293882.1 | - | 10 | 1396 | ||
| AF293883.1 | - | 10 | 1393 | |||
| AK148060.1 | - | 10 | 1457 | |||
| AK149933.1 | - | 10 | 1436 | |||
| AK167299.1 | - | 10 | 1410 | |||
| AK171226.1 | - | 10 | 1459 | |||
| AK171436.1 | - | 10 | 1436 | |||
| AK218322.1 | - | 2 | 94 | |||
| BC003298.1 | - | 6 | 754 | |||
| BC025493.1 | - | 10 | 1429 | |||

Pxn
Genomic Map