siRNA Details
| Gene Symbol: Rab12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024448.2 | NP_077768.2 | 4, 5 | 614 | ||
| GenBank mRNA | AB232607.1 | - | 4, 5 | 514 | ||
| AK013690.1 | - | 3, 4 | 270 | |||
| AK202686.1 | - | 2, 3 | 133 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab12| Gene Symbol: Rab12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024448.2 | NP_077768.2 | 4, 5 | 614 | ||
| GenBank mRNA | AB232607.1 | - | 4, 5 | 514 | ||
| AK013690.1 | - | 3, 4 | 270 | |||
| AK202686.1 | - | 2, 3 | 133 | |||