siRNA Details
| Gene Symbol: Rab23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159729.1 | NP_001153201.1 | 7 | 850 | ||
| NM_008999.4 | NP_033025.2 | 7 | 962 | |||
| GenBank mRNA | AB232618.1 | - | 5 | 527 | ||
| AK016493.1 | - | 7 | 862 | |||
| AK019666.1 | - | 4 | 411 | |||
| AK047068.1 | - | 7 | 963 | |||
| AK082017.1 | - | 7 | 926 | |||
| AK164057.1 | - | 7 | 1046 | |||
| BC025578.1 | - | 7 | 857 | 5323566 | ||
| Z22821.1 | - | 5 | 547 | |||

Rab23
Genomic Map